Product Details

SNP ID
rs148963507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37782781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACGTATTAAGTACCTACTGTTTG[C/T]TGGTGACTATGTAAACTAGAAAGGA
Phenotype
MIM: 606489
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC3 PubMed Links

Gene Details

Gene
EXOSC3
Gene Name
exosome component 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002269.2 Intron NP_001002269.1
NM_016042.3 Intron NP_057126.2
Gene
LOC105376037
Gene Name
uncharacterized LOC105376037
There are no transcripts associated with this gene.

Gene
TRMT10B
Gene Name
tRNA methyltransferase 10B
There are no transcripts associated with this gene.

View Full Product Details