Product Details

SNP ID
rs149117865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128833567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGCATGTGGGGATGTCAGGGCC[A/G]AGGCGTGACTTCAGGGCTAGAGTTC
Phenotype
MIM: 600547
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KYAT1 PubMed Links

Gene Details

Gene
KYAT1
Gene Name
kynurenine aminotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122671.1 1479 UTR 3 NP_001116143.1
NM_001122672.1 1479 UTR 3 NP_001116144.1
NM_001287390.1 1479 UTR 3 NP_001274319.1
NM_004059.4 1479 UTR 3 NP_004050.3
XM_006717311.3 1479 UTR 3 XP_006717374.1
XM_011519165.2 1479 UTR 3 XP_011517467.1
XM_011519167.2 1479 UTR 3 XP_011517469.2
XM_011519168.1 1479 UTR 3 XP_011517470.1
XM_011519169.1 1479 UTR 3 XP_011517471.1
XM_011519170.2 1479 UTR 3 XP_011517472.1
XM_017015260.1 1479 UTR 3 XP_016870749.1
XM_017015261.1 1479 UTR 3 XP_016870750.1
XM_017015262.1 1479 UTR 3 XP_016870751.1
XM_017015263.1 1479 UTR 3 XP_016870752.1
XM_017015264.1 1479 UTR 3 XP_016870753.1
XM_017015265.1 1479 UTR 3 XP_016870754.1
XM_017015266.1 1479 UTR 3 XP_016870755.1
XM_017015267.1 1479 UTR 3 XP_016870756.1
XM_017015268.1 1479 UTR 3 XP_016870757.1
Gene
SPOUT1
Gene Name
SPOUT domain containing methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016390.3 1479 Intron NP_057474.2
XM_017014804.1 1479 Intron XP_016870293.1

View Full Product Details