Product Details

SNP ID
rs149367725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124946654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTCTATAGGTTCACAAATTGGC[A/G]TTGCACGTATTTACTATCCTCTTAT
Phenotype
MIM: 602502
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLGA1 PubMed Links

Gene Details

Gene
GOLGA1
Gene Name
golgin A1
There are no transcripts associated with this gene.

Gene
SCAI
Gene Name
suppressor of cancer cell invasion
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144877.2 8165 UTR 3 NP_001138349.1
NM_173690.4 8165 UTR 3 NP_775961.2

View Full Product Details