Product Details

SNP ID
rs150075957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128706920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCTGAGCTGCTGGCGGAGGAGC[G/T]ACAGCATCGACTGCACGTTGAGGCA
Phenotype
MIM: 610714
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PKN3 PubMed Links

Gene Details

Gene
PKN3
Gene Name
protein kinase N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317926.1 1031 Missense Mutation CGA,CTA R183L NP_001304855.1
NM_013355.4 1031 Missense Mutation CGA,CTA R183L NP_037487.2
XM_005251946.3 1031 Missense Mutation CGA,CTA R180L XP_005252003.1
XM_006717080.2 1031 Missense Mutation CGA,CTA R190L XP_006717143.1
XM_017014649.1 1031 Missense Mutation CGA,CTA R190L XP_016870138.1
XM_017014650.1 1031 Missense Mutation CGA,CTA R57L XP_016870139.1

View Full Product Details