Product Details

SNP ID
rs150217084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128721771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGGCAGTGGTGGTCGTAGCGG[C/T]GGACGCAACGGCGGCACTCACGGCA
Phenotype
MIM: 610714
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100506100 PubMed Links

Gene Details

Gene
LOC100506100
Gene Name
uncharacterized LOC100506100
There are no transcripts associated with this gene.

Gene
PKN3
Gene Name
protein kinase N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317926.1 572 Intron NP_001304855.1
NM_013355.4 572 Intron NP_037487.2
XM_005251946.3 572 Intron XP_005252003.1
XM_006717080.2 572 Intron XP_006717143.1
XM_017014649.1 572 Intron XP_016870138.1
XM_017014650.1 572 Intron XP_016870139.1
Gene
ZDHHC12
Gene Name
zinc finger DHHC-type containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318015.1 572 Missense Mutation CAC,CGC H176R NP_001304944.1
NM_001318016.1 572 Missense Mutation CAC,CGC H121R NP_001304945.1
NM_001318020.1 572 Missense Mutation CAC,CGC H121R NP_001304949.1
NM_001318023.1 572 Missense Mutation CAC,CGC H120R NP_001304952.1
NM_032799.4 572 Missense Mutation CAC,CGC H121R NP_116188.2
XM_011519116.2 572 Missense Mutation CAC,CGC H176R XP_011517418.1
Gene
ZER1
Gene Name
zyg-11 related cell cycle regulator
There are no transcripts associated with this gene.

View Full Product Details