Product Details

SNP ID
rs150308649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129081143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTCCGGGTAAGATGGCAGCGGA[C/T]GGACAGTGCTCGCTCCCCGCTTCAT
Phenotype
MIM: 614516 MIM: 616774
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOLPP1 PubMed Links

Gene Details

Gene
DOLPP1
Gene Name
dolichyldiphosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135917.1 40 Silent Mutation GAC,GAT D4D NP_001129389.1
NM_020438.4 40 Silent Mutation GAC,GAT D4D NP_065171.2
Gene
MIGA2
Gene Name
mitoguardin 2
There are no transcripts associated with this gene.

View Full Product Details