Product Details

SNP ID
rs150529893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129635327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGAGGGCGTGATTCTGGACGAC[A/G]TGGACAGCAGCGTGTGCCGGGACCT
Phenotype
MIM: 615051 MIM: 613560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB6 PubMed Links

Gene Details

Gene
ASB6
Gene Name
ankyrin repeat and SOCS box containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202403.1 758 UTR 3 NP_001189332.1
NM_017873.3 758 UTR 3 NP_060343.1
NM_177999.2 758 UTR 3 NP_821066.1
Gene
NTMT1
Gene Name
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286796.1 758 Missense Mutation ATG,GTG M179V NP_001273725.1
NM_001286797.1 758 Missense Mutation ATG,GTG M179V NP_001273726.1
NM_001286798.1 758 Missense Mutation ATG,GTG M179V NP_001273727.1
NM_001286799.1 758 Missense Mutation ATG,GTG M179V NP_001273728.1
NM_001286800.1 758 UTR 3 NP_001273729.1
NM_001286801.1 758 UTR 3 NP_001273730.1
NM_001286802.1 758 Missense Mutation ATG,GTG M91V NP_001273731.1
NM_001286803.1 758 Missense Mutation ATG,GTG M91V NP_001273732.1
NM_014064.3 758 Missense Mutation ATG,GTG M179V NP_054783.2
XM_005251939.3 758 Missense Mutation ATG,GTG M91V XP_005251996.1
XM_017014642.1 758 Intron XP_016870131.1

View Full Product Details