Product Details

SNP ID
rs150545553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:86946039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGCTGCCGGGGCCGTTGCCCAG[C/T]TCGGCGCCGAAGCACAGGCGGGCCA
Phenotype
MIM: 139185
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAS1 PubMed Links

Gene Details

Gene
GAS1
Gene Name
growth arrest specific 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002048.2 1151 Silent Mutation GAA,GAG E247E NP_002039.2
Gene
GAS1RR
Gene Name
GAS1 adjacent regulatory RNA
There are no transcripts associated with this gene.

View Full Product Details