Product Details

SNP ID
rs150724884
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:38395767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAGCATGCTGCGCTTCCTGGCA[C/G]CCCGGCTGCTTAGCCTCCAGGGCAG
Phenotype
MIM: 100670
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ALDH1B1 PubMed Links

Gene Details

Gene
ALDH1B1
Gene Name
aldehyde dehydrogenase 1 family member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000692.4 172 Missense Mutation CCC,GCC P7A NP_000683.3
XM_011517802.2 172 Missense Mutation CCC,GCC P7A XP_011516104.1

View Full Product Details