Product Details

SNP ID
rs150784835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127455019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGCAAAAGAAGCTGGGGCAGAT[A/G]ACATTCTCGACATCTCTAAATGTGA
Phenotype
MIM: 610933 MIM: 180475 MIM: 194552
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRSAM1 PubMed Links

Gene Details

Gene
LRSAM1
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005373.3 1465 Missense Mutation AAC,GAC N32D NP_001005373.1
NM_001005374.3 1465 Missense Mutation AAC,GAC N32D NP_001005374.1
NM_001190723.2 1465 Missense Mutation AAC,GAC N32D NP_001177652.1
NM_138361.5 1465 Missense Mutation AAC,GAC N32D NP_612370.3
XM_006717316.3 1465 Missense Mutation AAC,GAC N32D XP_006717379.1
XM_017015283.1 1465 Missense Mutation AAC,GAC N32D XP_016870772.1
XM_017015284.1 1465 UTR 5 XP_016870773.1
Gene
RPL12
Gene Name
ribosomal protein L12
There are no transcripts associated with this gene.

Gene
SNORA65
Gene Name
small nucleolar RNA, H/ACA box 65
There are no transcripts associated with this gene.

Gene
ZNF79
Gene Name
zinc finger protein 79
There are no transcripts associated with this gene.

View Full Product Details