Product Details

SNP ID
rs151168466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104535928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAATTTTTCTTCCCCAATCAGGT[C/G]TTGAGACTTCGGTTTCGCATACATA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR13C3 PubMed Links

Gene Details

Gene
OR13C3
Gene Name
olfactory receptor family 13 subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001961.1 886 Missense Mutation CAC,GAC H296D NP_001001961.1
Gene
OR13C4
Gene Name
olfactory receptor family 13 subfamily C member 4
There are no transcripts associated with this gene.

View Full Product Details