Product Details

SNP ID
rs151206237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122159977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGCTAAAAACAGGCGCCATGG[A/G]GTACACAGGGAGCAAATATATCGGG
Phenotype
MIM: 603359
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MORN5 PubMed Links

Gene Details

Gene
MORN5
Gene Name
MORN repeat containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286828.1 66 Missense Mutation GAG,GGG E2G NP_001273757.1
NM_198469.3 66 Missense Mutation GAG,GGG E2G NP_940871.2
XM_005251877.4 66 Missense Mutation GAG,GGG E2G XP_005251934.1
XM_005251878.3 66 Intron XP_005251935.1
XM_006717034.3 66 Missense Mutation GAG,GGG E2G XP_006717097.1
XM_011518457.2 66 Missense Mutation GAG,GGG E2G XP_011516759.1
XM_011518458.1 66 UTR 5 XP_011516760.1
XM_017014578.1 66 Missense Mutation GAG,GGG E101G XP_016870067.1
Gene
NDUFA8
Gene Name
NADH:ubiquinone oxidoreductase subunit A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318195.1 66 Intron NP_001305124.1
NM_014222.2 66 Intron NP_055037.1

View Full Product Details