Product Details

SNP ID
rs111452972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15283584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTGATGACATGCTCGACCGAGA[C/T]ACTTCCGGACACACAGGCGGCAGAG
Phenotype
MIM: 300626
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASB11 PubMed Links

Gene Details

Gene
ASB11
Gene Name
ankyrin repeat and SOCS box containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012428.2 861 Missense Mutation TAT,TGT Y277C NP_001012428.1
NM_001201583.1 861 Missense Mutation TAT,TGT Y281C NP_001188512.1
NM_080873.2 861 Missense Mutation TAT,TGT Y298C NP_543149.1

View Full Product Details