Product Details

SNP ID
rs122463168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:137567454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAAGCAGGAGCTGTCGTGCAAG[G/T]GGATCGACGAGGCTCAGCTGAGCCG
Phenotype
MIM: 300265
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZIC3 PubMed Links

Gene Details

Gene
ZIC3
Gene Name
Zic family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003413.3 1268 Missense Mutation GGG,TGG G255W NP_003404.1
XM_017029802.1 1268 Missense Mutation GGG,TGG G255W XP_016885291.1

View Full Product Details