Product Details

SNP ID
rs138067286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101837792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCACACACTTACAGGGGTCTTT[C/G]AGAATTTTTATATTCCTGCTATCCT
Phenotype
MIM: 300319
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NXF5 PubMed Links

Gene Details

Gene
NXF5
Gene Name
nuclear RNA export factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032946.2 1269 Silent Mutation CTC,CTG L303L NP_116564.2

View Full Product Details