Product Details

SNP ID
rs138167528
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102553553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGACTGGTGACAAATTTGCTTA[A/G]CTTTTGTTTGCATTAAAATGTCACA
Phenotype
MIM: 300318
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NXF4 PubMed Links

Gene Details

Gene
NXF4
Gene Name
nuclear RNA export factor 4 pseudogene
There are no transcripts associated with this gene.

View Full Product Details