Product Details
- SNP ID
-
rs138276898
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:16738958 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGAGAACTAAGAAGTGTCCTTGG[G/T]GTTGGAGAACCAAGAGGTTTTGGTC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SYAP1
PubMed Links
Gene Details
- Gene
- SYAP1
- Gene Name
- synapse associated protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032796.3 |
|
Intron |
|
|
NP_116185.2 |
View Full Product Details