Product Details

SNP ID
rs138385391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:133026484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATACACTGATGTTTCCAGAAGT[C/T]ATCTTTCGAATAACTTTTAATAATT
Phenotype
MIM: 300309
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP26 PubMed Links

Gene Details

Gene
USP26
Gene Name
ubiquitin specific peptidase 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031907.1 1875 Missense Mutation ATA,ATG I579M NP_114113.1
XM_017029892.1 1875 Missense Mutation ATA,ATG I579M XP_016885381.1

View Full Product Details