Product Details

SNP ID
rs138947666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:139955925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTTAGGGTTACTTCAGAGGTGA[C/G]TTGATTGTACCCTCTGTCACACGTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CXorf66 PubMed Links

Gene Details

Gene
CXorf66
Gene Name
chromosome X open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013403.2 1079 Missense Mutation CTC,GTC L353V NP_001013421.1

View Full Product Details