Product Details

SNP ID
rs138953994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:10026654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCCAGCTAAGGCATTCTCACT[C/T]GTGTGTTTTCTGTTCCTTGCCGCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WWC3 PubMed Links

Gene Details

Gene
WWC3
Gene Name
WWC family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015691.3 Intron NP_056506.2

View Full Product Details