Product Details
- SNP ID
-
rs139220923
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:103079422 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGCTGAGTTTTAGGCAACGCACT[A/G]AGGGAGTCCACAATATCAAGTTTTG
- Phenotype
-
MIM: 300316
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NXF3
PubMed Links
Gene Details
- Gene
- NXF3
- Gene Name
- nuclear RNA export factor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022052.1 |
1343 |
Silent Mutation |
CTC,CTT |
L424L |
NP_071335.1 |
View Full Product Details