Product Details

SNP ID
rs139941345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:149884360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACCAGGGCTTATGGATGTGCAG[A/G]TTCCCACAGCTGAGGAGCAGAAGGC
Phenotype
MIM: 300892 MIM: 300341
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00850 PubMed Links

Gene Details

Gene
LINC00850
Gene Name
long intergenic non-protein coding RNA 850
There are no transcripts associated with this gene.

Gene
MAGEA8
Gene Name
MAGE family member A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166400.1 609 Missense Mutation ATT,GTT I30V NP_001159872.1
NM_001166401.1 609 Missense Mutation ATT,GTT I30V NP_001159873.1
NM_005364.4 609 Missense Mutation ATT,GTT I30V NP_005355.2
XM_017029520.1 609 Missense Mutation ATT,GTT I30V XP_016885009.1
Gene
MAGEA8-AS1
Gene Name
MAGEA8 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details