Product Details

SNP ID
rs140017010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:64188342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCCTCTCCTACAGGTGATTAAT[A/G]AGAGGTTGAGCTTGGCAAGATTAGC
Phenotype
MIM: 300647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMER1 PubMed Links

Gene Details

Gene
AMER1
Gene Name
APC membrane recruitment protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152424.3 5218 UTR 3 NP_689637.3

View Full Product Details