Product Details

SNP ID
rs140339263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54750204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGAAATCCAGAGGGATTTAAA[A/G]GAGGAAGAAGGAGGGGCAGCAAAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITIH6 PubMed Links

Gene Details

Gene
ITIH6
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198510.2 Intron NP_940912.1

View Full Product Details