Product Details

SNP ID
rs140500732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50192443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTGATTATTGACAATTACTTCA[C/T]GTCCAAGAGAGTCTTGATAGTTCTG
Phenotype
MIM: 300185
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKAP4 PubMed Links

Gene Details

Gene
AKAP4
Gene Name
A-kinase anchoring protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003886.2 2394 Missense Mutation CAT,CGT H757R NP_003877.2
NM_139289.1 2394 Missense Mutation CAT,CGT H748R NP_647450.1

View Full Product Details