Product Details
- SNP ID
-
rs140590028
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:103676831 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCCTCACGGATCCTGAAGGGGGG[C/T]TCTCTGCAGAGCGACCACCCCATCT
- Phenotype
-
MIM: 300409
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MORF4L2
PubMed Links
Gene Details
- Gene
- MORF4L2
- Gene Name
- mortality factor 4 like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001142418.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135890.1 |
NM_001142419.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135891.1 |
NM_001142420.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135892.1 |
NM_001142421.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135893.1 |
NM_001142422.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135894.1 |
NM_001142423.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135895.1 |
NM_001142424.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135896.1 |
NM_001142425.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135897.1 |
NM_001142426.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135898.1 |
NM_001142427.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135899.1 |
NM_001142428.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135900.1 |
NM_001142429.1 |
701 |
Intron |
|
|
NP_001135901.1 |
NM_001142430.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135902.1 |
NM_001142431.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135903.1 |
NM_001142432.1 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_001135904.1 |
NM_012286.2 |
701 |
Missense Mutation |
AAC,AGC |
N66S |
NP_036418.1 |
View Full Product Details