Product Details

SNP ID
rs140603964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47977683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTTTCCAAACTCATGACAGTCA[C/T]GAGCACTCTTGGTGATAATTGTTTT
Phenotype
MIM: 314993
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF182 PubMed Links

Gene Details

Gene
ZNF182
Gene Name
zinc finger protein 182
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007088.1 656 Missense Mutation CAT,CGT H116R NP_001007089.1
NM_001178099.1 656 Missense Mutation CAT,CGT H135R NP_001171570.1
NM_006962.1 656 Missense Mutation CAT,CGT H135R NP_008893.1

View Full Product Details