Product Details

SNP ID
rs140608748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:129739249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACTGGGGCTGCTGCCCCTGGCT[A/G]GTCCTCCTCTGTGGTATGTGCATCC
Phenotype
MIM: 300145
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XPNPEP2 PubMed Links

Gene Details

Gene
XPNPEP2
Gene Name
X-prolyl aminopeptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003399.5 280 Silent Mutation CTA,CTG L12L NP_003390.4

View Full Product Details