Product Details

SNP ID
rs141277897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15283509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCCAGGAACACTTAGGACTAT[C/T]GGTATAGGAGGAATCGTTCGAGTGG
Phenotype
MIM: 300626
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASB11 PubMed Links

Gene Details

Gene
ASB11
Gene Name
ankyrin repeat and SOCS box containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012428.2 936 Missense Mutation CAA,CGA Q302R NP_001012428.1
NM_001201583.1 936 Missense Mutation CAA,CGA Q306R NP_001188512.1
NM_080873.2 936 Missense Mutation CAA,CGA Q323R NP_543149.1

View Full Product Details