Product Details

SNP ID
rs141603294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71293283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGATATTCGTGATTCTGGTACA[C/T]ACCTAGCAAAAATTAGACTGAGCAT
Phenotype
MIM: 300332 MIM: 300084
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGB1BP2 PubMed Links

Gene Details

Gene
ITGB1BP2
Gene Name
integrin subunit beta 1 binding protein 2
There are no transcripts associated with this gene.

Gene
NONO
Gene Name
non-POU domain containing, octamer-binding
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145408.1 Intron NP_001138880.1
NM_001145409.1 Intron NP_001138881.1
NM_001145410.1 Intron NP_001138882.1
NM_007363.4 Intron NP_031389.3

View Full Product Details