Product Details

SNP ID
rs141647599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48773432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGTGTCCAACTACATCTCCTC[A/G]TGATGGAGGCTGGACCTCCTCCATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLOD5 PubMed Links

Gene Details

Gene
GLOD5
Gene Name
glyoxalase domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080489.2 624 Silent Mutation TCA,TCG S160S NP_001073958.2
XM_011543911.2 624 Silent Mutation TCA,TCG S118S XP_011542213.1
XM_011543912.2 624 Silent Mutation TCA,TCG S183S XP_011542214.1

View Full Product Details