Product Details

SNP ID
rs141781891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:85107395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCTTATTTGAAAGTAATCCATG[C/T]TTGTTTGCCTCATGCCCATTTGGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SATL1 PubMed Links

Gene Details

Gene
SATL1
Gene Name
spermidine/spermine N1-acetyl transferase-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012980.2 1574 Missense Mutation AAC,AGC N525S NP_001012998.2
XM_011530940.2 1574 Missense Mutation AAC,AGC N525S XP_011529242.1

View Full Product Details