Product Details

SNP ID
rs141889429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152167026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATGGCCTCTAAATAATATATTA[G/T]AAAATTGGCAATAAAGTTAATGATC
Phenotype
MIM: 305660
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GABRA3 PubMed Links

Gene Details

Gene
GABRA3
Gene Name
gamma-aminobutyric acid type A receptor alpha3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000808.3 2709 Intron NP_000799.1
XM_006724811.2 2709 UTR 3 XP_006724874.1

View Full Product Details