Product Details

SNP ID
rs142332875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:2949408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTCTCATCAGAAAGCAATCGGC[A/T]TGGACAATCAGAGCACCCACAAAAT
Phenotype
MIM: 300180
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARSE PubMed Links

Gene Details

Gene
ARSE
Gene Name
arylsulfatase E (chondrodysplasia punctata 1)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000047.2 770 Intron NP_000038.2
NM_001282628.1 770 Intron NP_001269557.1
NM_001282631.1 770 Missense Mutation CAA,CAT Q205H NP_001269560.1
XM_005274518.2 770 Intron XP_005274575.1
XM_005274519.4 770 Intron XP_005274576.1
XM_005274521.4 770 Intron XP_005274578.1
XM_011545521.1 770 Intron XP_011543823.1
XM_017029525.1 770 Intron XP_016885014.1
XM_017029526.1 770 Intron XP_016885015.1

View Full Product Details