Product Details

SNP ID
rs142803076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:69505813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTCCCTTTCCCGTGCCCCGGC[A/C]GAGTTCCCCTCCGCCAAAAAAAACT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM155B PubMed Links

Gene Details

Gene
FAM155B
Gene Name
family with sequence similarity 155 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015686.2 579 Silent Mutation GCA,GCC A177A NP_056501.2
XM_011530908.2 579 Silent Mutation GCA,GCC A177A XP_011529210.1
XM_011530909.2 579 Silent Mutation GCA,GCC A177A XP_011529211.1

View Full Product Details