Product Details

SNP ID
rs143297000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50192674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACTGGTACAGTCAAGTTCTTGA[C/T]GCTCCTGGCATTGTTCTTCCGCTTT
Phenotype
MIM: 300185
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKAP4 PubMed Links

Gene Details

Gene
AKAP4
Gene Name
A-kinase anchoring protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003886.2 2163 Missense Mutation CAT,CGT H680R NP_003877.2
NM_139289.1 2163 Missense Mutation CAT,CGT H671R NP_647450.1

View Full Product Details