Product Details

SNP ID
rs143532957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:41108288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCTCTCTGTTAAACCTCTCTCT[A/G]TGTTGGTATCACATGCACATATTAG
Phenotype
MIM: 300072
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP9X PubMed Links

Gene Details

Gene
USP9X
Gene Name
ubiquitin specific peptidase 9, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039590.2 Intron NP_001034679.2
NM_001039591.2 Intron NP_001034680.2
XM_005272675.4 Intron XP_005272732.1
XM_005272676.4 Intron XP_005272733.1

View Full Product Details