Product Details

SNP ID
rs143775522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:2935184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCACAAAGTGGACTTTCCACATT[G/T]TTCCTCCTGGTGGAAAGATAATCAT
Phenotype
MIM: 300002 MIM: 300180
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARSD PubMed Links

Gene Details

Gene
ARSD
Gene Name
arylsulfatase D
There are no transcripts associated with this gene.

Gene
ARSE
Gene Name
arylsulfatase E (chondrodysplasia punctata 1)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000047.2 1438 Intron NP_000038.2
NM_001282628.1 1438 Intron NP_001269557.1
NM_001282631.1 1438 Missense Mutation AAA,ACA K428T NP_001269560.1
XM_005274518.2 1438 Intron XP_005274575.1
XM_005274519.4 1438 Intron XP_005274576.1
XM_005274521.4 1438 Intron XP_005274578.1
XM_011545521.1 1438 Intron XP_011543823.1
XM_017029525.1 1438 Intron XP_016885014.1
XM_017029526.1 1438 Intron XP_016885015.1

View Full Product Details