Product Details

SNP ID
rs144037435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23335182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGTCATCGTCACCTCCTTCCAG[A/G]AAGCCAACATGCTGGACCAGCATCA
Phenotype
MIM: 300828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTCHD1 PubMed Links

Gene Details

Gene
PTCHD1
Gene Name
patched domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173495.2 315 Missense Mutation AAA,GAA K103E NP_775766.2
XM_011545449.2 315 Missense Mutation AAA,GAA K103E XP_011543751.1

View Full Product Details