Product Details

SNP ID
rs144541262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154400119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCTTCCTGCAGATACAAACAAA[A/G]CATGAGTAAGTCTTAGCAAGCTCTT
Phenotype
MIM: 300081 MIM: 312173
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNASE1L1 PubMed Links

Gene Details

Gene
DNASE1L1
Gene Name
deoxyribonuclease I-like 1
There are no transcripts associated with this gene.

Gene
LOC107987332
Gene Name
uncharacterized LOC107987332
There are no transcripts associated with this gene.

Gene
RPL10
Gene Name
ribosomal protein L10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256577.2 Intron NP_001243506.2
NM_001256580.2 Intron NP_001243509.2
NM_001303624.1 Intron NP_001290553.1
NM_001303625.1 Intron NP_001290554.1
NM_001303626.1 Intron NP_001290555.1
NM_006013.4 Intron NP_006004.3
Gene
SNORA70
Gene Name
small nucleolar RNA, H/ACA box 70
There are no transcripts associated with this gene.

View Full Product Details