Product Details

SNP ID
rs144948393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:9029370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTGCTCGTCTGTGATGTATTC[C/T]TCAAGGACATTTAGCAACTTCAGAG
Phenotype
MIM: 300478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM9B PubMed Links

Gene Details

Gene
FAM9B
Gene Name
family with sequence similarity 9 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205849.3 1268 Silent Mutation GAA,GAG E110E NP_995321.1
XM_011545464.2 1268 Silent Mutation GAA,GAG E110E XP_011543766.1
XM_011545465.2 1268 Intron XP_011543767.1

View Full Product Details