Product Details

SNP ID
rs144993154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:117903323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCTTGTATCTCTCTACTGACTT[C/T]CTTTTCCACTTACTTTAGCTTTGAA
Phenotype
MIM: 300655
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL13 PubMed Links

Gene Details

Gene
KLHL13
Gene Name
kelch like family member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168299.1 Intron NP_001161771.1
NM_001168300.1 Intron NP_001161772.1
NM_001168301.1 Intron NP_001161773.1
NM_001168302.1 Intron NP_001161774.1
NM_001168303.1 Intron NP_001161775.1
NM_033495.3 Intron NP_277030.2
XM_011531409.2 Intron XP_011529711.1
XM_011531410.2 Intron XP_011529712.1
XM_011531411.1 Intron XP_011529713.1
XM_017029950.1 Intron XP_016885439.1

View Full Product Details