Product Details

SNP ID
rs145029490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48914218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGGCTGGTGTTTGCATCCAGGG[A/G]TCCAGCAGGATCTCTTCCAGTGAGG
Phenotype
MIM: 300713 MIM: 300295 MIM: 314375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTUD5 PubMed Links

Gene Details

Gene
OTUD5
Gene Name
OTU deubiquitinase 5
There are no transcripts associated with this gene.

Gene
PIM2
Gene Name
Pim-2 proto-oncogene, serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006875.3 1151 Silent Mutation GAC,GAT D283D NP_006866.2
Gene
SLC35A2
Gene Name
solute carrier family 35 member A2
There are no transcripts associated with this gene.

View Full Product Details