Product Details

SNP ID
rs145052423
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:141905702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCAGTCTCCTCTCCAGAATTCT[C/T]AGAGTTCTCCTGAGGGGAAGGACTC
Phenotype
MIM: 300223 MIM: 300469
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MAGEC1 PubMed Links

Gene Details

Gene
MAGEC1
Gene Name
MAGE family member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005462.4 622 Missense Mutation CAG,TAG Q100* NP_005453.2
XM_011531418.2 622 Missense Mutation CAG,TAG Q100* XP_011529720.1
Gene
MAGEC3
Gene Name
MAGE family member C3
There are no transcripts associated with this gene.

View Full Product Details