Product Details

SNP ID
rs145306319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:43950693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGATTTATGTAATTGCATCCGCA[A/T]CTTCAGAGTCATTTGCGGCAAATAT
Phenotype
MIM: 300658
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NDP PubMed Links

Gene Details

Gene
NDP
Gene Name
NDP, norrin cystine knot growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000266.3 Intron NP_000257.1

View Full Product Details