Product Details

SNP ID
rs145695346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:133026245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAGTGAGGTATCTTCCAGATAC[A/G]TCATTAAGTGAGCTAACGGTTCTTT
Phenotype
MIM: 300309
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP26 PubMed Links

Gene Details

Gene
USP26
Gene Name
ubiquitin specific peptidase 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031907.1 2114 Missense Mutation ACG,ATG T659M NP_114113.1
XM_017029892.1 2114 Missense Mutation ACG,ATG T659M XP_016885381.1

View Full Product Details