Product Details

SNP ID
rs145731331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72301819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTCGGTCCAACCCCAGTTCCAC[C/T]ACCAGCGACATCAGCACTTCCTCAT
Phenotype
MIM: 300149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CITED1 PubMed Links

Gene Details

Gene
CITED1
Gene Name
Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144885.1 1062 Silent Mutation GTA,GTG V188V NP_001138357.1
NM_001144886.1 1062 Silent Mutation GTA,GTG V162V NP_001138358.1
NM_001144887.1 1062 Silent Mutation GTA,GTG V162V NP_001138359.1
NM_004143.3 1062 Silent Mutation GTA,GTG V162V NP_004134.2
XM_011530958.1 1062 Silent Mutation GTA,GTG V188V XP_011529260.1

View Full Product Details