Product Details
- SNP ID
-
rs145927426
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:134788775 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAAGGTGCTGGAGAAACTGGAGT[G/T]AAGTCAATTCTCTTAGGAGAATATA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FAM122B
PubMed Links
Gene Details
- Gene
- FAM122B
- Gene Name
- family with sequence similarity 122B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001166599.2 |
1408 |
Missense Mutation |
TTA,TTC |
L111F |
NP_001160071.1 |
NM_001166600.2 |
1408 |
Missense Mutation |
TTA,TTC |
L130F |
NP_001160072.1 |
NM_001170756.1 |
1408 |
Missense Mutation |
TTA,TTC |
L130F |
NP_001164227.1 |
NM_001170757.1 |
1408 |
Missense Mutation |
TTA,TTC |
L77F |
NP_001164228.1 |
NM_145284.5 |
1408 |
Missense Mutation |
TTA,TTC |
L111F |
NP_660327.2 |
XM_006724730.2 |
1408 |
Missense Mutation |
TTA,TTC |
L130F |
XP_006724793.1 |
XM_006724732.2 |
1408 |
Missense Mutation |
TTA,TTC |
L130F |
XP_006724795.1 |
XM_006724733.3 |
1408 |
Missense Mutation |
TTA,TTC |
L111F |
XP_006724796.1 |
XM_011531282.1 |
1408 |
Missense Mutation |
TTA,TTC |
L134F |
XP_011529584.1 |
XM_011531283.1 |
1408 |
Missense Mutation |
TTA,TTC |
L134F |
XP_011529585.1 |
XM_011531284.1 |
1408 |
Missense Mutation |
TTA,TTC |
L130F |
XP_011529586.1 |
XM_011531285.1 |
1408 |
Missense Mutation |
TTA,TTC |
L130F |
XP_011529587.1 |
XM_011531286.1 |
1408 |
Missense Mutation |
TTA,TTC |
L115F |
XP_011529588.1 |
XM_011531287.1 |
1408 |
Missense Mutation |
TTA,TTC |
L134F |
XP_011529589.1 |
XM_011531288.1 |
1408 |
Missense Mutation |
TTA,TTC |
L111F |
XP_011529590.1 |
XM_011531289.1 |
1408 |
Missense Mutation |
TTA,TTC |
L134F |
XP_011529591.1 |
XM_011531290.1 |
1408 |
Missense Mutation |
TTA,TTC |
L111F |
XP_011529592.1 |
XM_011531291.1 |
1408 |
Missense Mutation |
TTA,TTC |
L134F |
XP_011529593.1 |
XM_011531292.1 |
1408 |
Missense Mutation |
TTA,TTC |
L134F |
XP_011529594.1 |
XM_011531293.1 |
1408 |
Missense Mutation |
TTA,TTC |
L130F |
XP_011529595.1 |
XM_011531294.1 |
1408 |
Missense Mutation |
TTA,TTC |
L111F |
XP_011529596.1 |
XM_017029306.1 |
1408 |
Missense Mutation |
TTA,TTC |
L115F |
XP_016884795.1 |
XM_017029307.1 |
1408 |
Missense Mutation |
TTA,TTC |
L115F |
XP_016884796.1 |
- Gene
- FAM122C
- Gene Name
- family with sequence similarity 122C
There are no transcripts associated with this gene.
View Full Product Details