Product Details

SNP ID
rs146249944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30242721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCAGAGCAGTGCCTGGACCCTT[C/T]CAAGGAATGGGCTTCTGATGCCTCT
Phenotype
MIM: 300097 MIM: 300152 MIM: 300153
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAGEB1 PubMed Links

Gene Details

Gene
MAGEB1
Gene Name
MAGE family member B1
There are no transcripts associated with this gene.

Gene
MAGEB3
Gene Name
MAGE family member B3
There are no transcripts associated with this gene.

Gene
MAGEB4
Gene Name
MAGE family member B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002367.3 782 Missense Mutation CCA,TCA P196S NP_002358.1

View Full Product Details