Product Details

SNP ID
rs146590513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130631440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCATTGTACCCAGGCATTGTA[C/T]GTGGCATGTGTGCATTAGCTTCCTT
Phenotype
MIM: 300282
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENOX2 PubMed Links

Gene Details

Gene
ENOX2
Gene Name
ecto-NOX disulfide-thiol exchanger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281736.1 Intron NP_001268665.1
NM_006375.3 Intron NP_006366.2
NM_182314.2 Intron NP_872114.1
XM_005262354.3 Intron XP_005262411.1
XM_011531244.2 Intron XP_011529546.1
XM_011531245.2 Intron XP_011529547.1
XM_011531247.2 Intron XP_011529549.1
XM_011531249.2 Intron XP_011529551.1
XM_011531251.2 Intron XP_011529553.1
XM_017029226.1 Intron XP_016884715.1
XM_017029227.1 Intron XP_016884716.1
XM_017029228.1 Intron XP_016884717.1
XM_017029229.1 Intron XP_016884718.1

View Full Product Details